Anti-MMP1 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:50~100 For IF starting dilution is: 1:10~50
Type: Primary
Antigen: MMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Size:400 ul
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:54 kDa
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Epitope:Amino Acid Range = 317-347
- Form:Liquid
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- ImmunoFluorescence:Yes
- Host:Rabbit
- Concentration:0.35 mg/mL
- Cat. No.:76011-284
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:MMP1
- Gene ID:P03956
- Immunogen:This MMP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 317-347 amino acids from the Central region of human MMP1.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:MMP1
- Antigen Name:Interstitial collagenase
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.



