Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PDE4B Rabbit Polyclonal Antibody
Anti-PDE4B Rabbit Polyclonal Antibody
Catalog # 76010-948
Supplier:  Prosci
undefined
Anti-PDE4B Rabbit Polyclonal Antibody
Catalog # 76010-948
Supplier:  Prosci
Supplier Number:  55-986

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    cAMP-specific 3',5'-cyclic phosphodiesterase 4B
  • Antigen Symbol:
    PDE4B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 ul
  • Epitope:
    Amino Acid Range = 189-217
  • Form:
    Liquid
  • Gene ID:
    Q07343
  • Antigen Synonyms:
    PDE4B
  • Modification:
    None
  • Storage Buffer:
    Supplied in PBS with 0.09% (W/V) sodium azide.
  • Molecular Weight:
    83 kDa
  • Storage Temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0.26 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This PDE4B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 189-217 amino acids from the Central region of human PDE4B.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Cat. No.:
    76010-948

Specifications

About this item

This gene is a member of the type IV, cyclic AMP (cAMP)-specific, cyclic nucleotide phosphodiesterase (PDE) family. Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. The cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. This gene encodes a protein that specifically hydrolyzes cAMP. Altered activity of this protein has been associated with schizophrenia and bipolar affective disorder. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: PDE4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human