Anti-SLC16A8 Rabbit Polyclonal Antibody
Catalog # 76010-902
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Monocarboxylate transporter 3
- Antigen Symbol:SLC16A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 466-493
- Form:Liquid
- Gene ID:O95907
- Antigen Synonyms:SLC16A8
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:52 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.4 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This SLC16A8 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 466-493 amino acids from the C-terminal region of human SLC16A8.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76010-902
Specifications
About this item
SLC16A8 is a member of a family of proton-coupled monocarboxylate transporters that mediate lactate transport across cell membranes (Yoon et al., 1999 [PubMed 10493836]).[supplied by OMIM].
For WB starting dilution is: 1:1000
Type: Primary
Antigen: SLC16A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human