Anti-LDLRAD3 Rabbit Polyclonal Antibody
Catalog # 76010-758
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Low-density lipoprotein receptor class A domain-containing protein 3
- Antigen Symbol:LDLRAD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 317-345
- Form:Liquid
- Gene ID:Q86YD5
- Antigen Synonyms:LDLRAD3
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:37 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.27 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This LDLRAD3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 317-345 amino acids from the C-terminal region of human LDLRAD3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76010-758
Specifications
About this item
The specific function of the protein remains unknown.
For WB starting dilution is: 1:1000
Type: Primary
Antigen: LDLRAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse