- Antibody Type:Primary
- Antigen Name:Histone H2A.Z
- Antigen Symbol:H2AFZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 59-86
- Form:Liquid
- Gene ID:P0C0S5
- Antigen Synonyms:H2AFZ
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:14 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This H2AFZ antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 59-86 amino acids from the C-terminal region of human H2AFZ.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76010-336
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene encodes a replication-independent member of the histone H2A family that is distinct from other members of the family. Studies in mice have shown that this particular histone is required for embryonic development and indicate that lack of functional histone H2A leads to embryonic lethality. [provided by RefSeq].
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: H2AFZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human