Specifications
- Antibody Type:Primary
- Antigen Name:Otoancorin
- Antigen Symbol:OTOA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 92-120
- Form:Liquid
- Gene ID:Q7RTW8
- Antigen Synonyms:OTOA
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:129 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This OTOA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 92-120 amino acids from the N-terminal region of human OTOA.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76010-170
Specifications
About this item
The protein encoded by this gene is specifically expressed in the inner ear, and is located at the interface between the apical surface of the inner ear sensory epithelia and their overlying acellular gels. It is prposed that this protein is involved in the attachment of the inner ear acellular gels to the apical surface of the underlying nonsensory cells. Mutations in this gene are associated with autosomal recessive deafness type 22 (DFNB22). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq].
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: OTOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse