Anti-METAP1D Rabbit Polyclonal Antibody
Catalog # 76009-188
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Methionine aminopeptidase 1D, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03174}
- Antigen Symbol:METAP1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 306-334
- Form:Liquid
- Gene ID:Q6UB28
- Antigen Synonyms:METAP1D
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:37 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This MAP1D antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 306-334 amino acids from the N-terminal region of human MAP1D.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76009-188
Specifications
About this item
The N-terminal methionine excision pathway is an essential process in which the N-terminal methionine is removed from many proteins, thus facilitating subsequent protein modification. In mitochondria, enzymes that catalyze this reaction are celled methionine aminopeptidases (MetAps, or MAPs; EC 3.4.11.18) (Serero et al., 2003 [PubMed 14532271]).
For WB starting dilution is: 1:1000
Type: Primary
Antigen: METAP1D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse