Anti-IGF2BP1 Rabbit Polyclonal Antibody
Catalog # 76009-460
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Insulin-like growth factor 2 mRNA-binding protein 1
- Antigen Symbol:IGF2BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 508-534
- Form:Liquid
- Gene ID:Q9NZI8
- Antigen Synonyms:IGF2BP1
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:63 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This IGF2BP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 508-534 amino acids from the C-terminal region of human IGF2BP1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76009-460
Specifications
About this item
IGF2BP1 is a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein encoded by this gene contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene.
For FACS starting dilution is: 1:25 For WB starting dilution is: 1:2000
Type: Primary
Antigen: IGF2BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse