Specifications
- Antibody Type:Primary
- Antigen Name:Probable ATP-dependent RNA helicase DDX11
- Antigen Symbol:DDX11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 656-683
- Form:Liquid
- Gene ID:Q96FC9
- Antigen Synonyms:DDX11
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:108 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This DDX11 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 656-683 amino acids from the Central region of human DDX11.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76009-444
Specifications
About this item
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX11 encodes a DEAD box protein, which is an enzyme that possesses both ATPase and DNA helicase activities. DDX11 is a homolog of the yeast CHL1 gene, and may function to maintain chromosome transmission fidelity and genome stability.
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: DDX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse