Anti-SLC5A12 Rabbit Polyclonal Antibody
76008-844
: Prosci
- Antibody Type:Primary
- Antigen Name:Sodium-coupled monocarboxylate transporter 2
- Antigen Symbol:SLC5A12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 576-605
- Form:Liquid
- Gene ID:Q1EHB4
- Antigen Synonyms:SLC5A12
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:68 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.5 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This SLC5A12 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 576-605 amino acids from the C-terminal region of human SLC5A12.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76008-844
Normal blood lactate is maintained at about 1.5 mM, and little filtered lactate is excreted in urine. Reabsorption of lactate is mediated by the low-affinity Na(+)-coupled lactate transporter SLC5A12 in the initial part of the proximal tubule and by the high-affinity Na(+)-coupled lactate transporter SLC5A8 (MIM 608044) in the distal proximal tubule (Gopal et al., 2007 [PubMed 17692818]).
For WB starting dilution is: 1:2000 For IHC-P starting dilution is: 1:50~100 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: SLC5A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human