Anti-CLDN22 Rabbit Polyclonal Antibody
Catalog # 76009-294
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Claudin-22
- Antigen Symbol:CLDN22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 ul
- Epitope:Amino Acid Range = 90-117
- Form:Liquid
- Gene ID:Q8N7P3
- Antigen Synonyms:CLDN22
- Modification:None
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Molecular Weight:25 kDa
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0.43 mg/mL
- Shipping Temperature:4 °C
- Immunogen:This CLDN22 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 90-117 amino acids from the Central region of human CLDN22.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:76009-294
Specifications
About this item
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. This gene is intronless and overlaps the 3' UTR of the WWC2 gene (GeneID: 80014) on the opposite strand.
For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: CLDN22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse