Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-FGF9 Rabbit Polyclonal Antibody
Catalog # 76009-034
Supplier:  Prosci
undefined
Anti-FGF9 Rabbit Polyclonal Antibody
Catalog # 76009-034
Supplier:  Prosci
Supplier Number:  55-089

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Fibroblast growth factor 9
  • Antigen Symbol:
    FGF9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    400 ul
  • Epitope:
    Amino Acid Range = 31-58
  • Form:
    Liquid
  • Gene ID:
    P31371
  • Antigen Synonyms:
    FGF9
  • Modification:
    None
  • Storage Buffer:
    Supplied in PBS with 0.09% (W/V) sodium azide.
  • Molecular Weight:
    23 kDa
  • Storage Temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This FGF9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 31-58 amino acids from the N-terminal region of human FGF9.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification.
  • Cat. No.:
    76009-034

Specifications

About this item

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.

For WB starting dilution is: 1:1000 For FACS starting dilution is: 1:10~50

Type: Primary
Antigen: FGF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse