Anti-PRMT3 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Arginine methylation is an irreversible post translational modification which has only recently been linked to protein activity. At least three types of PRMT enzymes have been identified in mammalian cells. These enzymes have been shown to have essential regulatory functions by methylation of key proteins in several fundamental areas. These protein include nuclear proteins, IL enhancer binding factor, nuclear factors, cell cycle proteins, signal transduction proteins, apoptosis proteins, and viral proteins. The mammalian PRMT family currently consists of 7 members that share two large domains of homology. Outside of these domains, epitopes were identified and antibodies against all 7 PRMT members have been developed.
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: PRMT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Size:400 ul
- Clonality:Polyclonal
- Modification:None
- Molecular Weight:60 kDa
- Purification:This antibody is purified through a protein G column, followed by dialysis against PBS.
- Epitope:Amino Acid Range = 142-173
- Form:Liquid
- Storage Buffer:Supplied in PBS with 0.09% (W/V) sodium azide.
- Host:Rabbit
- Cat. No.:76008-898
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:PRMT3
- Gene ID:O60678
- Immunogen:This PRMT3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 142-173 amino acids from the N-terminal region of human PRMT3.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:PRMT3
- Antigen Name:Protein arginine N-methyltransferase 3
- Storage Temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



