HardyDisks™ Nitrate HardyDisks, Hardy Diagnostics
Nitrate, HardyDisk, 1×50 disks
Catalog # 75993-562
Supplier: Hardy Diagnostics
Supplier #: Z7071
HardyDisks™ Nitrate HardyDisks, Hardy Diagnostics
Catalog # 75993-562
Supplier: Hardy Diagnostics
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Documents
Specifications
Specifications
- Description:Nitrate, HardyDisk
- Size:1×50 disks