- Description:Cefotetan, CTT-30, HardyDisks
- Size:5×50 disks
- Cat. No.:75993-458
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
- Impregnated disks
- Spring loaded dispenser
- Smooth and reliable dispensing
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.