Order Entry
Puerto Rico
ContactUsLinkComponent
Actinomycin D (synthetic) ≥95% (by HPLC)
Actinomycin D (synthetic) ≥95% (by HPLC)
Catalog # 102989-160
Supplier:  Adipogen
CAS Number:  
Actinomycin D (synthetic) ≥95% (by HPLC)
Catalog # 102989-160
Supplier:  Adipogen
Supplier Number:  CDX-A0043-M020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Size:
    20 mg
  • Cat. No.:
    102989-160
  • Supplier no.:
    CDXA0043M020

Specifications

About this item

 :
 :1255.5 g/mol
Storage Temperature:Freezer
UN:3462
ADR:6.1,III
CAS Number:50-76-0
Formula:C₆₂H₈₆N₁₂O₁₆
MDL Number:MFCD00005033