Gram's iodine solution stabilized for QuickSlide instrumentation
Catalog # 75993-542
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Dropping bottle, brown
- Size:100 ml (4 oz.)
- Cat. No.:75993-542
Specifications
About this item
:
:419.812 g/mol
CAS Number:12298-68-9
Formula:I₃K
MDL Number:MFCD00134128