Order Entry
United States
ContactUsLinkComponent
Brady® BBP®85 and PowerMark® Industrial Grade Print Ribbon
Brady® BBP®85 and PowerMark® Industrial Grade Print Ribbon
Catalog # 10841-222
Supplier:  Brady Worldwide
undefined
Brady® BBP®85 and PowerMark® Industrial Grade Print Ribbon
Catalog # 10841-222
Supplier:  Brady Worldwide
Supplier Number:  13511

Specifications

  • Color:
    Black
  • Description:
    BBP®85 and powerMark® industrial grade print ribbon
  • Length:
    61 m (200 ft.)
  • Environmentally Preferable:
  • Cat. No.:
    10841-222
  • Width:
    158.7 mm (6.25")
  • Supplier no.:
    13511

Specifications

About this item

This industrial-grade ribbon is uniquely designed with memory cells that communicate information to the BBP®85 and PowerMark® sign and label printers.

BBP®85 and PowerMark® Industrial Grade Print Ribbon is super-tough and smear-proof to ensure printed text lasts for years, both outdoors and in industrial areas.