ContactUsLinkComponent
BBL™ Brucella Broth, BD Biosciences
BBL™ Brucella Broth, BD Biosciences
Catalog # 90000-076
Supplier:  BD
CAS Number:  
BBL™ Brucella Broth, BD Biosciences
Catalog # 90000-076
Supplier:  BD
Supplier Number:  211088
CAS Number:  
Specifications
  • Description:
    Brucella Broth
  • Size:
    500 g
  • Cat. No.:
    90000-076
  • Supplier no.:
    211088
Specifications
About this item
Brucella Broth is used for the cultivation of Brucella species and for the isolation and cultivation of a wide variety of fastidious and nonfastidious microorganisms.
  • Use for the isolation and cultivation of a wide variety of microorganisms
  • Isolates and cultivates nutritionally fastidious specimens
  • Can also be used to cultivate Campylobacter
This medium is recommended for the cultivation of Brucella species and was recommended as one Brucella Media of several media suitable for use as the liquid medium component of biphasic blood culture bottles.