Order Entry
Puerto Rico
ContactUsLinkComponent
 
Alcohol Oxidase (from P. pastoris), MP Biomedicals
  IC02190155.2
 :  
Alcohol Oxidase (from P. pastoris), MP Biomedicals
  IC02190155.2
 :  02190155.2
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • CAS No.:
    9073-63-6
  • Source:
    P. pastoris
  • Size:
    250 units
  • Storage Conditions:
    –20 °C
  • MDL No.:
    MFCD00130452
  • Enzyme Name:
    Alcohol oxidase
  • Molecular Weight:
    ~630000
  • Cat. No.:
    IC02190155.2

 

 

Alcohol oxidase has been detected in several strains of yeast,such as Candida, Pichia, and Hansenula,that utilize methanol as a sole carbon and energy source

One unit will oxidize 1.0 µmol of methanol to formaldehyde per minute at pH 7.5 at 25 °C.

Some of these enzymes have been isolated and partially characterized. Alcohol oxidase plays a major role in the metabolism of methanol resulting in the formation of formaldehyde. Primarily localized in the peroxisome, alcohol oxidase has also been found in the cytoplasm. Monomers are synthesized in the cytosol and assembled into octomers in the peroxisome. Octomerization is thought to be chaparone mediated. Alcohol oxidase is of interest for the study of protein translocation into peroxisomes. It catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide.