Order Entry
Puerto Rico
ContactUsLinkComponent
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Irritant
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Catalog # IC0219024705
Supplier:  MP Biomedicals
CAS Number:  
Irritant
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Catalog # IC0219024705
Supplier:  MP Biomedicals
Supplier Number:  0219024705
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Size:
    5 g
  • Cat. No.:
    IC0219024705

Specifications

Specification Test Results

  • Physical Appearance:
    White Powder
  • Vapor Pressure:
    3.01E-07 mmHg at 25 °C (lit.)
  • Optical Rotation:
    (c=2, Ethanol) +59° to +62°
  • Solubility:
    Soluble in water (3 mg/mL) at 25 °C, PBS pH 7.2, SOL., ethanol, DMSO, DMF (100 mg/mL), chloroform, dichloromethane, ether, ethyl acetate, methanol, and THF. Soluble in acetone (50 mg/mL).

Specification Test Results

About this item

CAS Number:22204-53-1
 :
Formula:C₁₄H₁₄O₃
MDL Number:MFCD00010500
 :230.26 g/mol
Boiling Pt:403.9 °C (760 mmHg)) (lit.)
Boiling Pt:403,9 °C (760 mmHg)) (lit.)
Melting Pt:152…160 °C
Density:1,20 g/cm³ (lit.)
Storage Temperature:Ambient
EINECS:244-838-7
UN:2811
ADR:6.1,III
 :GHS07