Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND

Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND

EDTA, 0.5M, 100 mL, 8.0
Catalog # RLMB-014
Supplier #:  MB-014
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND
Catalog # RLMB-014
CAS Number:  
Buffer, EDTA Buffer Solution pH 8,0 (0,5 mol/L), ROCKLAND

EDTA, 0.5M, 100 mL, 8.0

CAS Number:  
Price
$123.44
Log in for availability
Each (100ml)
Minimum order quantity is 1
Product Details & Documents

EDTA is widely used for scavenging metal ion in biochemistry and molecular biology

In tissue culture EDTA is used as a chelating agent that binds to calcium and prevents joining of cadherins between cells, preventing clumping of cells grown in liquid suspension, or detaching adherent cells for passaging. EDTA is also known to inhibit a range of metallopeptidases, the method of inhibition occurs via the chelation of the metal ion required for catalytic activity.

Ethylenediaminetetraacetic acid is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer consists of 0.5 M Trisodium EDTA at pH 8.0. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water (DI) treated with diethyl pyrocarbonate (DEPC).

Ion depletion is commonly used to deactivate metal-dependent enzymes, either as an assay for their reactivity or to suppress damage to DNA or proteins

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.

Documents

Specifications

Specifications

  • Description:
    EDTA, 0.5M
  • Size:
    100 mL
  • pH:
    8.0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.