About this item
High quality silver nanoparticles. Provided with full COA for great value. Displaceable citrate surface.
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- Standard colloidal concentration (0.02 mg/mL, 20 ppm, 0.185 mM, OD's of 0.8 to 2.7 [depending on size])
- Citrate surface is stable and displaceable
Spherical silver nanoparticles, 0.02 mg/mL
Each batch is extensively characterized to ensure it meets the highest quality standards, providing reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.