ContactUsLinkComponent
Fiber Optics Voice and Data Kit
Fiber Optics Voice and Data Kit
Catalog # 470227-036
Supplier:  Elenco Electronics
undefined
Fiber Optics Voice and Data Kit
Catalog # 470227-036
Supplier:  Elenco Electronics
Supplier Number:  FO30K
Specifications
  • Description:
    Fiber Optics Voice and Data Kit
  • Cat. No.:
    470227-036
Specifications
About this item
Learn the basics of fiber optics communications while you build this exciting kit.
  • Includes instruction manual
  • Includes 3 foot fiber optic cable
Transmit your voice or a 1kHz signal through the fiber optic cable to a fiber optic receiver. Comes complete with all of the necessary components; including PC boards; microphone; speaker and fiber optic cable. Transmitter and receiver (BOTH included) on separate boards; can be located hundreds of feet apart. Use for science projects.