Order Entry
United States
ContactUsLinkComponent
Digital Temperature Controlled Solder Station
Digital Temperature Controlled Solder Station
Catalog # 470227-030
Supplier:  Elenco Electronics
undefined
Digital Temperature Controlled Solder Station
Catalog # 470227-030
Supplier:  Elenco Electronics
Supplier Number:  SL30A

Specifications

  • Description:
    Digital temperature controlled solder station
  • Environmentally Preferable:
  • Cat. No.:
    470227-030

Specifications

About this item

Professional Soldering Station 48W (160-480 C or 320-900 F) with a 42 inch long cable.

  • Heater Power for a 48 watt Soldering Iron
  • Iron Voltage is 24 VAC (station runs on 110-130 VAC 60Hz)

The digital display is switchable to read out in C or F.