Air Flow Indicator Kit, Sensidyne
Catalog # 89091-766
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Description:Smoke Tube Air Flow Indicator Kit (HVAC)
- Cat. No.:89091-766
- Supplier no.:7014094
Specifications
About this item
This Air Flow Indicator (HVAC) Kit is highly recommended to track air flow currents and for fit testing respirators per OSHA 29 CFR part 1910.134, irritant fume protocol.
Delivery: The kit includes carrying pouch, aspirator bulb, tube tip breaker, and a box of smoke tubes.