ContactUsLinkComponent
SDS-Page Running Gel Buffers, Rockland
  RLMB-018
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
SDS-Page Running Gel Buffers, Rockland
  RLMB-018
 :  MB-018
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    SDS-PAGE Sample Buffer without DTT or [beta]MeOH, 2X
  • Size:
    100 mL
  • Environmentally Preferable:
  • Cat. No.:
    RLMB-018
  • pH:
    6.8
  • Supplier no.:
    MB-018
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature
SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins
 : This product is for research use only and is not intended for therapeutic or diagnostic applications.