Order Entry
Puerto Rico
ContactUsLinkComponent
Competent Cell Buffer, Rockland Immunochemicals
Competent Cell Buffer, Rockland Immunochemicals
Catalog # RLMB-057
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Competent Cell Buffer, Rockland Immunochemicals
Catalog # RLMB-057
Supplier Number:  MB-057
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    Competent Cell Buffer, 60mM CaCl2, 15% Glycerol, 10mM TRIS HCl
  • Size:
    100 mL
  • Cat. No.:
    RLMB-057
  • pH:
    7.5
  • Supplier no.:
    MB-057

Specifications

About this item

This product is ready-to-use as a working 1X solution and requires no further dilution

Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.

This buffer consists of 60 mM Calcium Chloride, 15% v/v Glycerol, 10mM Tris HCl at a pH of 7.5

This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.