ContactUsLinkComponent
ELAN Coolant Fluid, PerkinElmer
ELAN Coolant Fluid, PerkinElmer
  97012-606
undefined
ELAN Coolant Fluid, PerkinElmer
  97012-606
 :  WE016558
  • Capacity:
    1 L
  • Description:
    Coolant Fluid for ELAN
  • Cat. No.:
    97012-606
  • Supplier no.:
    WE016558
Coolant fluid for use with NexION and ELAN 9000/6x00/DRC inductively coupled plasma mass spectrometry (ICP-MS) models.
  • Available as 1 litre bottle