Order Entry
Puerto Rico
ContactUsLinkComponent
Accessories for Gibson Assembly® HiFi HC Cloning Kits, SGI-DNA
Accessories for Gibson Assembly® HiFi HC Cloning Kits, SGI-DNA
Catalog # 75997-590
Supplier:  TELESIS BIO
CAS Number:  
Accessories for Gibson Assembly® HiFi HC Cloning Kits, SGI-DNA
Catalog # 75997-590
Supplier:  TELESIS BIO
Supplier Number:  GA1000-05C
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Gibson Assembly® Positive control
  • Cat. No.:
    75997-590
  • No. of reactions:
    5

Specifications

About this item