Open-Head UN Rated Poly Drum, With Metal Lever-Lock Ring, Eagle Mfg
Open-head poly drums, 55 gal., 10.7 kg (23.5 lbs.), 21", 36⅜", Blue, Metal lever locking ring, 1H2/X200/S, 1H2/Y250/S
Catalog # 75845-054
Supplier: Eagle Manufacturing
Supplier #: 1656MB
Open-Head UN Rated Poly Drum, With Metal Lever-Lock Ring, Eagle Mfg
Catalog # 75845-054
Supplier: Eagle Manufacturing
Jump to:
Product Details & Documents
Drum is constructed of blue blow-molded high-density polyethylene (HDPE) with UV inhibitors and has a white lid with a metal lever-lock band closure.
- Durable
- Lightweight
- Chemical and weather resistant
Ideal for safe storage, handling, and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal, and incineration.
Certifications: Complies to DOT 46, CFR 173.12, and UN DOT Rating 1H2/X200/S, 1H2/Y250/S.
Documents
Eagle MFG Poly Drums
Product Literature
Specifications
- Ø:21"
- Capacity:55 gal.
- Color:Blue
- Description:Open-head poly drums
- Height:36⅜"
- Un markings:1H2/X200/S, 1H2/Y250/S
- Weight:10.7 kg (23.5 lbs.)
- Lid type:Metal lever locking ring
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



