WL Nutrient Agar, HiMedia
Supplier: HiMedia
Certificates
About this item
WL Nutrient Agar is recommended for selective isolation and enumeration of bacteria encountered in breweries and industrial fermentations.
Specifications
- Storage Conditions:2 - 8 °C
- Form:Light yellow to light green homogeneous free flowing powder
- Storage:<30 °C
- pH:5.30 - 5.70
Specifications
Related Information
Certificates
Search by Lot Number95020-592
SDS
F&BFOCUS Volume 5 Issue 1w
Product Literature
HiMedia WL Nutrient Agar M115
Datasheet
Frequently Bought Together