Order Entry
Puerto Rico
ContactUsLinkComponent
Lactosyl ceramides ≥99%
Lactosyl ceramides ≥99%
Catalog # 100291-312
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Lactosyl ceramides ≥99%
Catalog # 100291-312
Supplier Number:  5610721MG
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 mg
  • Cat. No.:
    100291-312
  • Supplier no.:
    5610721MG

Specifications

About this item

 :
 :960(C23:0) g/mol
UN:12352100
ADR:,
CAS Number:4682-48-8
MDL Number:MFCD00133445