Order Entry
Puerto Rico
ContactUsLinkComponent
Hexadecanoic acid-2-¹³C (Palmitic) ≥98%
Hexadecanoic acid-2-¹³C (Palmitic) ≥98%
Catalog # 100291-950
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Hexadecanoic acid-2-¹³C (Palmitic) ≥98%
Catalog # 100291-950
Supplier Number:  781602100MG
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 mg
  • Cat. No.:
    100291-950
  • Supplier no.:
    781602100MG

Specifications

About this item

 :
 :257.422 g/mol
ADR:,
CAS Number:287100-87-2
Formula:CH₃(CH₂)₁₃¹³CH₂CO₂H
MDL Number:MFCD00083983