About this item
Purified, concentrated, high quality gold nanoparticles. Sterile and endotoxin-free. PVP surface for stability.
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- High concentration materials (1 mg/mL, 5.08 mM, 1000 ppm, ~16 to 30 OD)
- Extensively purified to remove residual reactants
- Manufactured sterile; provided in ultra-pure (Milli-Q) DI water
- BioPure materials ideal for in-vitro and in-vivo experiments, nanotoxicology
- PVP surface provides strong steric stability
Spherical gold nanoparticles, 1 mg/mL (high concentration)
Each batch is extensively characterized to meet the highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.