Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lead Standard, 1000 ppm w/w (NIST) for AAS, Aqua Solutions
Catalog # 103193-788
Supplier:  Aqua Solutions
CAS Number:  
Lead Standard, 1000 ppm w/w (NIST) for AAS, Aqua Solutions
Catalog # 103193-788
Supplier:  Aqua Solutions
Supplier Number:  5250-100ML
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    3.5% HNO₃
  • Pack type:
    Clear Plastic
  • Size:
    100 ml
  • Cat. No.:
    103193-788
  • Element:
    Lead
  • Supplier no.:
    5250-100ML

Specifications

About this item