
(+)-Rutin trihydrate ≥95% (by NMR)
Catalog # 102988-162
Supplier: Adipogen
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Size:5 g
- Cat. No.:102988-162
- Supplier no.:AGCN20408G005
Specifications
Specification Test Results
- Source/Host:Isolated from Ruta graveolens.
- Purity:95% (NMR)
- Appearance/Color:Pale yellow powder.
- Solubility:Soluble in DMSO, acetone, dichloromethane or ethyl acetate.
Specification Test Results
About this item
:
:664.58 g/mol (610.52 anhy)
:GHS07
Storage Temperature:Refrigerator
CAS Number:250249-75-3
Formula:C₂₇H₃₀O₁₆·3H₂O
MDL Number:MFCD00149490