Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Filgotinib (GLPG0634) ≥95%

Filgotinib (GLPG0634) ≥95%

Marketsource
Supplier:   Adipogen
Filgotinib (GLPG0634) ≥95%
Supplier:  Adipogen
Marketsource
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

GLPG0634 is a novel and specific inhibitor of JAK1 and JAK2 with IC(50) of 10nM and 28nM respectively measured by in vitro kinase assays (ATP 2µM). In particular GLPG0634, while being a potent inhibitor of JAK1 and JAK2 does not inhibit TYK2 or JAK3, and GLPG0634 exhibits a dramatically improved in vivo potency over structurally similar compounds. May be used for the prevention of variety of conditions in animals including inflammatory conditions, autoimmune diseases, proliferative diseases, transplant rejection, diseases involving impairment of cartilage turnover, congenital cartilage malformation and/or diseases associated with hypersecretion of IL-6.

Synonyms:  N-(5-(4-((1,1-Dioxidothiomorpholino)methyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide, Filgotinib
  • CAS Number:1206161-97-8
  • Formula:C₂₁H₂₃N₅O₃S
  • MDL Number:MFCD20527867
  • Molecular Weight:425.5 g/mol
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Purity:
    95%
  • Appearance/Color:
    Solid.
  • Solubility:
    Soluble in DMSO.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic vial
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $283.85
    $283.85
    Each (1mg)
       $283.85

    Supplier Number: SYN-1158-M001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $479.08
    $479.08
    Each (5mg)
       $479.08

    Supplier Number: SYN-1158-M005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    10 mg
    Log in for availability
    Log in for availability
    Each (10mg)
    Each (10mg) - $688.15
    $688.15
    Each (10mg)
       $688.15

    Supplier Number: SYN-1158-M010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.