Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Afatinib ≥95%

Afatinib ≥95%

Marketsource
Supplier:   Adipogen
Afatinib ≥95%
Supplier:  Adipogen
Marketsource
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Afatinib is an irreversible kinase inhibitor and binds to the kinase domains of EGFR (ErbB1), HER2 (ErbB2), and HER4 (ErbB4) to inhibit tyrosine kinase autophosphorylation. This results in a downregulation of ErbB signaling and subsequent inhibition of proliferation of cell lines expressing wild-type EGFR, selected EGFR exon 19 deletion mutations, or exon 21 L858R mutations. It also inhibited in vitro proliferation of cell lines overexpressing HER2. Overall, tumor growth was inhibited by Afatinib very effectively with low nanomolar IC(50) values ranging from approximately 6nM to below 500nM.

Synonyms:  (S,E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-7-((tetrahydrofuran-3-yl)oxy)quinazolin-6-yl)-4-(dimethylamino)but-2-enamide, BIBW2992, Tovok
  • CAS Number:850140-72-6
  • Formula:C₂₄H₂₅ClFN₅O₃
  • MDL Number:MFCD12407405
  • Molecular Weight:485.94 g/mol
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Purity:
    >95%
  • Appearance/Color:
    Solid.
  • Solubility:
    Soluble in DMSO or ethanol.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic vial
    1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $119.08
    $119.08
    Each (1mg)
       $119.08

    Supplier Number: SYN-1100-M001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $180.00
    $180.00
    Each (5mg)
       $180.00

    Supplier Number: SYN-1100-M005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    10 mg
    Log in for availability
    Log in for availability
    Each (10mg)
    Each (10mg) - $299.08
    $299.08
    Each (10mg)
       $299.08

    Supplier Number: SYN-1100-M010
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Plastic vial
    50 mg
    Log in for availability
    Log in for availability
    Each (50mg)
    Each (50mg) - $822.46
    $822.46
    Each (50mg)
       $822.46

    Supplier Number: SYN-1100-M050
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.