About this item
Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome, calpains and other chymotrypsin-like proteases. Extinction: 380nm. Emission: 460 nm. This substrate is useful for inhibitor screening and kinetic analysis.
- Soluble in DMSO.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Source:
- Species:
- Environmentally Preferable: