Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
FSL-1

FSL-1

Vial, 100 µg
Catalog # 102988-458
Marketsource
Supplier:   Adipogen
Supplier #:  AG-CP3-0009-C100
FSL-1
Catalog # 102988-458
Supplier:  Adipogen
Marketsource
CAS Number:  322455-70-9
FSL-1

Vial, 100 µg

CAS Number:  322455-70-9
Price
$263.08
Log in for availability
Each (100µG)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

FSL-1 (Pam2CGDPKHPKSF) is a synthetic lipoprotein that represents the N-terminal part of the 44kDa lipoprotein LP44 of Mycoplasma salivarium. Stimulator of TLR2/TLR6. Inducer of TNF-alpha production in macrophages. Upregulates proinflammatory cytokines. Activator of the proinflammatory transcription factor NF-kappaB.

Synonyms:  Fibroblast-stimulating Lipopeptide
  • CAS Number:322455-70-9
  • Formula:C₈₄H₁₄₀N₁₄O₁₈S
  • Molecular Weight:1666.2 g/mol
  • Storage Temperature:Freezer
Specifications

Specification Test Results

  • Source/Host:
    Synthetic.
  • Purity:
    Absence of detectable protein or DNA contaminants with agonistic TLR activity.
  • Appearance/Color:
    Colorless powder.
  • Solubility:
    Soluble in water (10mg/ml). Use a homogeniser or sonicator to prepare a homogenous solution.
  • Formulation:
    Lyophilized.

Specifications

  • Pack type:
    Vial
  • Size:
    100 µg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.