Order Entry
Puerto Rico
ContactUsLinkComponent
Z-VRPR-FMK Trifluoroacetate Salt
Z-VRPR-FMK Trifluoroacetate Salt
Supplier:  Adipogen
Certificates
Z-VRPR-FMK Trifluoroacetate Salt
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    1381885-28-4 (free base)
  • Storage Conditions:
    Freezer
  • Form:
    White to slightly yellow solid
  • Protein Synonyms:
    Z-Val-Arg-Pro-DL-Arg-FMK
  • Protein/Peptide Name:
    Z-VRPR-FMK trifluoroacetate salt
  • Purity:
    ≥90% (by HPLC)
  • Solubility:
    Soluble in water
  • Molecular Weight:
    676.8
  • Molecular Formula:
    C₃₁H₄₉FN₁₀O₆·C₂HF₃O₂

Specifications