Convulxin
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Glycoprotein GPVI receptor agonist. Can be used for studies on platelet receptors. Activates mammalian platelets via binding and clustering of GPVI-receptors under physiological conditions. Occupation and clustering of GPVI activates Src family kinases, phosphorylates Fc receptor gamma-chain and activates p72SYK that is critical for downstream activation of platelets. Causes cardiovascular and respiratory disturbances. Prostaglandins are strong inhibitors of convulxin-induced aggregation. Heterodimeric C-type lectin. Disulfide linked heterodimer consisting of homologous alpha and beta chains. The heterodimers are additionally linked by disulfide bridges to form cyclic alpha(4)beta(4)heterotetramers.
- CAS Number:37206-04-5
- Molecular Weight:~84 kDa
- Storage Temperature:Freezer
- UN:3462
- ADR:6.1,I
Specifications
Specification Test Results
- Source/Host:Isolated from Crotalus durissus terrificus snake venom.
- Purity:Partially purified by multi-step chromatography.
- Appearance/Color:Lyophilized.
- Solubility:Soluble in organic buffer (HEPES).
Specifications
- Pack type:Vial
- Size:50 µg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



