Order Entry
Puerto Rico
ContactUsLinkComponent
Helenalin ≥97% (by HPLC)
Toxic
Helenalin ≥97% (by HPLC)
Catalog # 102988-266
Supplier:  Adipogen
CAS Number:  
Toxic
Helenalin ≥97% (by HPLC)
Catalog # 102988-266
Supplier:  Adipogen
Supplier Number:  AG-CN2-0435-C500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Size:
    500 µg
  • Cat. No.:
    102988-266
  • Supplier no.:
    AGCN20435C500

Specifications

About this item

 :
Storage Temperature:Freezer
UN:3462
ADR:6.1,III
 :GHS06
CAS Number:6754-13-8