Order Entry
Puerto Rico
ContactUsLinkComponent
Lactacystin ≥98% (by HPLC)
Lactacystin ≥98% (by HPLC)
Supplier:  Adipogen
Certificates
Lactacystin ≥98% (by HPLC)
Supplier:  Adipogen

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Source/Host:
    Isolated from Streptomyces lactacystinnaeus strain OM-6519.
  • Purity:
    98% (HPLC)
  • Appearance/Color:
    White to off-white solid.
  • Solubility:
    Soluble in water, methanol, ethanol, DMSO, pyridine, acetonitrile or dimethyl formamide. Insoluble in hexane, benzene, chloroform or ethyl acetate.

Specification Test Results