White Line Inducing Principle ≥98%
Catalog # 102987-226
Supplier: Adipogen
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Vial
- Size:1 mg
- Cat. No.:102987-226
- Supplier no.:AGCN20069M001
Specifications
Specification Test Results
- Source/Host:Isolated from Pseudomonas sp.
- Purity:98%
- Appearance/Color:White solid.
- Solubility:Soluble in DMSO, ethanol or methanol.
Specification Test Results
About this item
:
:1126.4 g/mol
Storage Temperature:Freezer
CAS Number:135096-89-8
Formula:C₅₄H₉₅N₉O₁₆