Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine dihydrochloride

3,3',5,5'-Tetramethylbenzidine dihydrochloride

Marketsource
Supplier:   Adipogen
3,3',5,5'-Tetramethylbenzidine dihydrochloride
Supplier:  Adipogen
Marketsource
Certificates
3,3',5,5'-Tetramethylbenzidine dihydrochloride

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TMBZ is a chromogenic reagent utilized for peroxidase detection. It has been developed as an alternative to benzidine. Although the TMBZ solution is colorless, it turns bluish-green (lambdamax: 655 nm) in the presence of hydrogen peroxide and peroxidase. The structure of this bluish-green complex is thought to be a radical form of two oxidized TMBZ molecules. This peroxidase substrate is used in staining procedures in immunohistochemistry as well as being a visualising reagent used in enzyme-linked immunosorbent assays (ELISA). The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2M H2SO4 (resulting in a yellow color), and read at 450nm.

Synonyms:  Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride
  • CAS Number:64285-73-0
  • Formula:C₁₆H₂₀N₂·2HCl
  • MDL Number:MFCD00012961
  • Molecular Weight:313.27 g/mol
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $81.69
    $81.69
    Each (1g)
       $81.69

    Supplier Number: CDX-T0151-G001
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • Vial
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $326.77
    $326.77
    Each (5g)
       $326.77

    Supplier Number: CDX-T0151-G005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.