Emodin
About this item
p56lck protein kinase inhibitor. Competitive casein kinase II (CK2) inhibitor. Anticancer agent with apoptosis inducing with antiangiogenic activity. Antioxidant, vasorelaxant and neuroprotective agent. NF-kappaB activation inhibitor. Inflammasome inhibitor. Potent selective 11beta-HSD1 inhibitor. Improves insulin sensitivity and lipid metabolism. Estrogen receptor ERalpha and ERbeta inhibitor. Inhibits androgen receptor (AR) nuclear translocation and induces AR degradation. VEGFR2, FGFR-1, EGFR, PDGFR-beta and Kit inhibitor. JAK2 inhibitor. Anti-diabetic agent. AMPK activator and PPARgamma agonist. Adipogenesis inhibitor. P2X7 receptor antagonist. Wnt signaling & PI3K/Akt/GSK-3beta pathway modulator.
- CAS Number:518-82-1
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00001207
- Molecular Weight:270.24 g/mol
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
p56lck protein kinase inhibitor. Competitive casein kinase II (CK2) inhibitor. Anticancer agent with apoptosis inducing with antiangiogenic activity. Antioxidant, vasorelaxant and neuroprotective agent. NF-kappaB activation inhibitor. Inflammasome inhibitor. Potent selective 11beta-HSD1 inhibitor. Improves insulin sensitivity and lipid metabolism. Estrogen receptor ERalpha and ERbeta inhibitor. Inhibits androgen receptor (AR) nuclear translocation and induces AR degradation. VEGFR2, FGFR-1, EGFR, PDGFR-beta and Kit inhibitor. JAK2 inhibitor. Anti-diabetic agent. AMPK activator and PPARgamma agonist. Adipogenesis inhibitor. P2X7 receptor antagonist. Wnt signaling & PI3K/Akt/GSK-3beta pathway modulator.
- CAS Number:518-82-1
- Formula:C₁₅H₁₀O₅
- MDL Number:MFCD00001207
- Molecular Weight:270.24 g/mol
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: AG-CN2-0457-M050Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- Supplier Number: AG-CN2-0457-M250Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



