BioProfile® FLEX is a chemistry/cell viability modular instrument that measures up to 15 key cell culture attributes.
- Saves as much as 30 minutes per sample compared to using multiple instruments
- Analysis as fast as 2 minutes per sample
Used for monitoring the progress of bioreactor, determining consumption and production of key metabolities, identification of growth-limiting nutrients, design of feeding strategies, calibration of bioreactor probes, measurement of cellular respiration, balancing media electrolytes and control levels of waste products.
By combining as many as five separate instruments into one easy-to-use instrument, BioProfile® FLEX simplifies workflow and saves time, labor, and operating cost versus multiple instruments.