Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tyloxapol

Tyloxapol

Irritant
Health
Tyloxapol
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tyloxapol is a liquid polymer that is nonionic and of the alkyl aryl polyether type. Tyloxapol is used as a surfactant, detergent, dispersing agent, encapsulating agent and hydroxy radical scavenger. It is a non-ionic liquid polymer that is soluble in polar and non-polar solvents.

GHS hazard : Exclamation mark;Health hazard

  • CAS Number:25301-02-4
  • MDL Number:MFCD00149002
  • Density:1,100
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $273.21
    $273.21
    Each (5g)
       $273.21

    Supplier Number: T1420-5GM
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $773.95
    $773.95
    Each (25g)
       $773.95

    Supplier Number: T1420-25GM
  • Drum
    20 kg
    Log in for availability
    Log in for availability
    Each (20kg)
    Each (20kg)

    Supplier Number: T1420-20KGBL

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.